4'-Chlor-3'-Fluoracetophenon
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-(4-Chloro-3-fluorophenyl)ethanone
PC302106-1G
PC302106-25G
PC302106-5G
APOSPC302106-1GEA
164
DKK
APOSPC302106-1G
APOSPC302106-25G
APOSPC302106-5G
4'-Chlor-3'-Fluoracetophenon
4'-Chlor-3'-Fluoracetophenon
Formel:
C₈H₆ClFO Molekylvægt: 172.59 g/mol Smeltepunkt: 39…42 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04115858 CAS-nummer: 151945-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...