α,α,α-Trifluor-3,5-dinitrotoluen 99%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3,5-Dinitrobenzotrifluoride
PC3018-100G
PC3018-25G
PC3018-500G
APOSPC3018-100GEA
523
DKK
APOSPC3018-100G
APOSPC3018-25G
APOSPC3018-500G
α,α,α-Trifluor-3,5-dinitrotoluen 99%
α,α,α-Trifluor-3,5-dinitrotoluen
Formel:
C₇H₃F₃N₂O₄ Molekylvægt: 236.11 g/mol Kogepunkt: 127…129 °C/9mmHg Smeltepunkt: 47…52 °C Flammepunkt: >110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007233 CAS-nummer: 401-99-0 EINECS: 206-935-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...