2,3-Dihydro-6-(trifluoromethyl)benzo[1,4]thiazin-3-one 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC2954E-100MG PC2954E-500MG
APOSPC2954E-100MGEA 305 DKK
APOSPC2954E-100MG APOSPC2954E-500MG
2,3-Dihydro-6-(trifluoromethyl)benzo[1,4]thiazin-3-one 97%
2,3-Dihydro-6-(trifluoromethyl)benzo[1,4]thiazin-3-one
Formel: C₉H₆F₃NOS
Smeltepunkt: 192…194 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00178754
CAS-nummer: 716-82-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR