Kalium(4-chlorphenyl)trifluorborat 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (4-Chlorophenyl)trifluoroboric acid potassium salt, Potassium (4-chlorophenyl)trifluoroborate

PC2917-10G PC2917-25G PC2917-5G
APOSPC2917-10GEA 1120 DKK
APOSPC2917-10G APOSPC2917-25G APOSPC2917-5G
Kalium(4-chlorphenyl)trifluorborat 95+%
Kalium(4-chlorphenyl)trifluorborat
Formel: C₆H₄BClF₃K
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04115770
CAS-nummer: 661465-44-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR