Fmoc-2-fluoro-DL-phenylglycine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-2-(2-fluorophenyl)acetic acid

PC27856-100MG PC27856-1G PC27856-250MG
APOSPC27856-100MGEA 3500 DKK
APOSPC27856-100MG APOSPC27856-1G APOSPC27856-250MG
Fmoc-2-fluoro-DL-phenylglycine
Fmoc-2-fluoro-DL-phenylglycine
Formel: C₂₃H₁₈FNO₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02682470
CAS-nummer: 433291-96-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR