4,4'-Difluorbenzophenon 98+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Bis(4-Fluorophenyl)methanone, Bis(4-fluorophenyl) ketone

PC2696-1KG PC2696-250G
APOSPC2696-1KGEA 818 DKK
APOSPC2696-1KG APOSPC2696-250G
4,4'-Difluorbenzophenon 98+%
4,4'-Difluorbenzophenon

Intermediate for the manufacture of polyetheretherketone(PEEK) high performance polymer.

Formel: C₁₃H₈F₂O
Molekylvægt: 218.2 g/mol
Kogepunkt: 170…172 °C/10mmHg
Smeltepunkt: 102…110 °C
Massefylde: 1.39 g/cm³ (25 °C)
Flammepunkt: 152 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000353
CAS-nummer: 345-92-6
EINECS: 206-466-3
UN nummer: 3077
ADR: 9+EHS III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR