4-Bromo-2-fluoro-5-nitroaniline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC2673-1G PC2673-5G PC2673-25G
APOSPC2673-1GEA 208 DKK
APOSPC2673-1G APOSPC2673-5G APOSPC2673-25G
4-Bromo-2-fluoro-5-nitroaniline
4-Bromo-2-fluoro-5-nitroaniline
Formel: C₆H₄BrFN₂O₂
Smeltepunkt: 92…94 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08741367
CAS-nummer: 87547-06-6
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR