2',3'-Difluoracetophenon 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-(2,3-Difluorophenyl)ethan-1-one, 1-(2,3-Difluorophenyl)ethanone, 2,3-Difluoroacetophenone

PC2574-100G PC2574-25G PC2574-5G
APOSPC2574-100GEA 2160 DKK
APOSPC2574-100G APOSPC2574-25G APOSPC2574-5G
2',3'-Difluoracetophenon 98%
2',3'-Difluoracetophenon
Formel: C₈H₆F₂O
Kogepunkt: 85 °C/20mmHg
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00061274
CAS-nummer: 18355-80-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR