2-Phenyl-5-(trifluoromethyl)oxazole-4-carboxylic acid 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC2355-1G PC2355-250MG
APOSPC2355-1GEA 819 DKK
APOSPC2355-1G APOSPC2355-250MG
2-Phenyl-5-(trifluoromethyl)oxazole-4-carboxylic acid 95+%
2-Phenyl-5-(trifluoromethyl)oxazole-4-carboxylic acid
Formel: C₁₁H₆F₃NO₃
Smeltepunkt: 170…172 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00203251
CAS-nummer: 236736-23-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR