5-Fluor-2-nitrobenzoesyre 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC2319-100G PC2319-25G PC2319-5G
APOSPC2319-100GEA 589 DKK
APOSPC2319-100G APOSPC2319-25G APOSPC2319-5G
5-Fluor-2-nitrobenzoesyre 98%
5-Fluor-2-nitrobenzoesyre
Formel: C₇H₄FNO₄
Smeltepunkt: 131…138 °C
Flammepunkt: >110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00055635
CAS-nummer: 320-98-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR