Methyl-3-amino-5-fluorobenzoate 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 3-amino-5-fluorobenzoate

PC201353-25G PC201353-5G
APOSPC201353-25GEA 2980 DKK
APOSPC201353-25G APOSPC201353-5G
Methyl-3-amino-5-fluorobenzoate 98%
Methyl-3-amino-5-fluorobenzoate
Formel: C₈H₈FNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05863683
CAS-nummer: 884497-46-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR