Methyl-4-amino-2-fluorobenzoate 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 4-amino-2-fluorobenzoate

PC201296-10G PC201296-100G PC201296-25G
APOSPC201296-10GEA 623 DKK
APOSPC201296-10G APOSPC201296-100G APOSPC201296-25G
Methyl-4-amino-2-fluorobenzoate 98%
Methyl-4-amino-2-fluorobenzoate
Formel: C₈H₈FNO₂
Molekylvægt: 169.1530 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09037917
CAS-nummer: 73792-08-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR