2-Fluoro-4-(4-methyl-1H-1,2,3-triazol-1-yl)aniline
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC200331-1G
PC200331-500MG
APOSPC200331-1GEA
1860
DKK
APOSPC200331-1G
APOSPC200331-500MG
2-Fluoro-4-(4-methyl-1H-1,2,3-triazol-1-yl)aniline
2-Fluoro-4-(4-methyl-1H-1,2,3-triazol-1-yl)aniline
Formel:
C₉H₉FN₄ Molekylvægt: 192.2 g/mol Smeltepunkt: 145…146 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD20921599 CAS-nummer: 1353878-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...