6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carbohydrazide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC200209-1G PC200209-250MG
APOSPC200209-1GEA 1050 DKK
APOSPC200209-1G APOSPC200209-250MG
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carbohydrazide
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carbohydrazide
Formel: C₉H₇F₃N₄O
Smeltepunkt: 202…203 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11100215
CAS-nummer: 1048912-65-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR