4-Chlor-3-Fluoranisol 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Chloro-2-fluoro-4-methoxybenzene
PC1824J-100G
PC1824J-25G
PC1824J-5G
APOSPC1824J-100GEA
1640
DKK
APOSPC1824J-100G
APOSPC1824J-25G
APOSPC1824J-5G
4-Chlor-3-Fluoranisol 98%
4-Chlor-3-Fluoranisol
Formel:
C₇H₆ClFO Kogepunkt: 200…204 °C Massefylde: 1,29 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00042570 CAS-nummer: 501-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...