4'-Chlor-2'-fluoracetanilid 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N-(4-Chloro-2-fluorophenyl)acetamide

PC1815-250G PC1815-50G
APOSPC1815-250GEA 1830 DKK
APOSPC1815-250G APOSPC1815-50G
4'-Chlor-2'-fluoracetanilid 98%
4'-Chlor-2'-fluoracetanilid
Formel: C₈H₇ClFNO
Smeltepunkt: 154…156 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00042520
CAS-nummer: 59280-70-5
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR