2-Chlor-1-Fluor-3-nitrobenzen

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Chloro-3-fluoronitrobenzene

PC1418-25G PC1418-5G
APOSPC1418-25GEA 1920 DKK
APOSPC1418-25G APOSPC1418-5G
2-Chlor-1-Fluor-3-nitrobenzen
2-Chlor-1-Fluor-3-nitrobenzen
Formel: C₆H₃ClFNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04112549
CAS-nummer: 21397-07-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR