2H-Nonafluorobutane 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC1286-100G PC1286-25G
APOSPC1286-100GEA 10720 DKK
APOSPC1286-100G APOSPC1286-25G
2H-Nonafluorobutane 97%
2H-Nonafluorobutane
Formel: C₄HF₉
Kogepunkt: 15 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03094114
CAS-nummer: 680-17-1
UN nummer: 3163
ADR: 2.2,

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR