2,6-Difluor-3-methoxybenzoesyre

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC1128-1G PC1128-250MG
APOSPC1128-1GEA 1310 DKK
APOSPC1128-1G APOSPC1128-250MG
2,6-Difluor-3-methoxybenzoesyre
2,6-Difluor-3-methoxybenzoesyre
Formel: C₈H₆F₂O₃
Molekylvægt: 188.13 g/mol
Smeltepunkt: 136…138 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04115901
CAS-nummer: 886498-30-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR