4-Nitro-2-(trifluormethyl)anilin 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Amino-5-nitro-α,α,α-trifluortoluene, 2-Amino-5-nitro-α,α,α-trifluorotoluene, 4-Nitro-2-trifluoromethylaniline, 2-Amino-5-nitrobenzotrifluoride, 4-Nitro-α,α,α-trifluoro-o-toluidine

PC1080-100G PC1080-10G PC1080-25G
APOSPC1080-100GEA 327 DKK
APOSPC1080-100G APOSPC1080-10G APOSPC1080-25G
4-Nitro-2-(trifluormethyl)anilin 95%
4-Nitro-2-(trifluormethyl)anilin
Formel: C₇H₅F₃N₂O₂
Smeltepunkt: 90…92 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007365
CAS-nummer: 121-01-7
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR