2-Bromo-5-(trifluoromethoxy)benzyl bromide 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
PC1067-1G
PC1067-5G
APOSPC1067-1GEA
985
DKK
APOSPC1067-1G
APOSPC1067-5G
2-Bromo-5-(trifluoromethoxy)benzyl bromide 95%
2-Bromo-5-(trifluoromethoxy)benzyl bromide
Formel:
C₈H₅Br₂F₃O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07368721 CAS-nummer: 886763-13-9 UN nummer: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...