4,5-Difluor-2-nitrobenzoesyre
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC1053-25G
PC1053-5G
PC1053-100G
APOSPC1053-25GEA
186
DKK
APOSPC1053-25G
APOSPC1053-5G
APOSPC1053-100G
4,5-Difluor-2-nitrobenzoesyre
4,5-Difluor-2-nitrobenzoesyre
Formel:
C₇H₃F₂NO₄ Smeltepunkt: 165…167 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00799521 CAS-nummer: 20372-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...