4-Brom-3-(trifluormethyl)anilin 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Amino-2-bromobenzotrifluoride, 4-Bromo-α,α,α-trifluoro-m-toluidine, 5-Amino-2-bromo-α,α,α-trifluorotoluene
PC1046-25G
PC1046-5G
PC1046-100G
APOSPC1046-25GEA
186
DKK
APOSPC1046-25G
APOSPC1046-5G
APOSPC1046-100G
4-Brom-3-(trifluormethyl)anilin 97%
4-Brom-3-(trifluormethyl)anilin
Formel:
C₇H₅BrF₃N Molekylvægt: 240.02 g/mol Kogepunkt: 81…84 °C/0.5mmHg Smeltepunkt: 46…49 °C Flammepunkt: >110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007827 CAS-nummer: 393-36-2 EINECS: 206-885-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...