Methyl-4-[(3-fluorobenzyl)oxy]benzenecarboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 4-[(3-fluorobenzyl)oxy]benzoate

PC10345-1G PC10345-250MG
APOSPC10345-1GEA 698 DKK
APOSPC10345-1G APOSPC10345-250MG
Methyl-4-[(3-fluorobenzyl)oxy]benzenecarboxylate
Methyl-4-[(3-fluorobenzyl)oxy]benzenecarboxylate
Formel: C₁₅H₁₃FO₃
Smeltepunkt: 71…73 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01568714
CAS-nummer: 454473-78-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR