α,α,α-Trifluor-m-toluidin 99%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
3-(Trifluoromethyl)aniline, 3-Aminobenzotrifluoride
PC1030-100G
PC1030-1KG
PC1030-500G
APOSPC1030-100GEA
164
DKK
APOSPC1030-100G
APOSPC1030-1KG
APOSPC1030-500G
α,α,α-Trifluor-m-toluidin 99%
α,α,α-Trifluor-m-toluidin
Intermediate for the synthesis of several pharmaceuticals including Bendrofluazide
Formel:
C₇H₆F₃N Kogepunkt: 187…189 °C Smeltepunkt: 44352 °C Massefylde: 1,295 g/cm³ (25 °C) Flammepunkt: 85 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007797 CAS-nummer: 98-16-8 EINECS: 202-643-4 UN nummer: 2948 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...