2-(2-Chloro-6-fluorobenzyl)malononitrile
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC10259-1G
PC10259-500MG
PC10259-5G
APOSPC10259-1GEA
2070
DKK
APOSPC10259-1G
APOSPC10259-500MG
APOSPC10259-5G
2-(2-Chloro-6-fluorobenzyl)malononitrile
2-(2-Chloro-6-fluorobenzyl)malononitrile
Formel:
C₁₀H₆ClFN₂ Molekylvægt: 208.62 g/mol Smeltepunkt: 132…134 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00231634 CAS-nummer: 338965-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...