1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanone
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4-Acetyl-5-chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole
PC10213-1G
PC10213-500MG
APOSPC10213-1GEA
5040
DKK
APOSPC10213-1G
APOSPC10213-500MG
1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanone
1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanone
Formel:
C₇H₆ClF₃N₂O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04117795 CAS-nummer: 863305-84-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...