3-{2-[3-(Trifluoromethyl)phenoxy]phenyl}acrylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC10207-1G PC10207-500MG PC10207-5G
APOSPC10207-1GEA 3050 DKK
APOSPC10207-1G APOSPC10207-500MG APOSPC10207-5G
3-{2-[3-(Trifluoromethyl)phenoxy]phenyl}acrylic acid
3-{2-[3-(Trifluoromethyl)phenoxy]phenyl}acrylic acid
Formel: C₁₆H₁₁F₃O₃
Smeltepunkt: 156…158 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01814780
CAS-nummer: 449778-81-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR