1-Brom-2-Fluor-4-nitrobenzen 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Bromo-3-fluoronitrobenzene

PC0996-25G PC0996-5G PC0996-100G
APOSPC0996-25GEA 317 DKK
APOSPC0996-25G APOSPC0996-5G APOSPC0996-100G
1-Brom-2-Fluor-4-nitrobenzen 97%
1-Brom-2-Fluor-4-nitrobenzen
Formel: C₆H₃BrFNO₂
Smeltepunkt: 88…94 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05865088
CAS-nummer: 185331-69-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR