2-Bromo-4-(trifluoromethyl)benzyl alcohol 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC0849-5G PC0849-25G
APOSPC0849-5GEA 844 DKK
APOSPC0849-5G APOSPC0849-25G
2-Bromo-4-(trifluoromethyl)benzyl alcohol 98%
2-Bromo-4-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆BrF₃O
Smeltepunkt: 60…61 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03788534
CAS-nummer: 497959-33-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR