5-Bromo-3-fluorosalicylaldehyde 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Bromo-3-fluoro-2-hydroxybenzaldehyde

PC0842-5G PC0842-1G PC0842-25G
APOSPC0842-5GEA 240 DKK
APOSPC0842-5G APOSPC0842-1G APOSPC0842-25G
5-Bromo-3-fluorosalicylaldehyde 97%
5-Bromo-3-fluorosalicylaldehyde
Formel: C₇H₄BrFO₂
Molekylvægt: 219.0079 g/mol
Smeltepunkt: 112…116 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03788530
CAS-nummer: 251300-28-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR