5-Fluor-2-nitroanisol 99%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Fluoro-2-methoxy-1-nitrobenzene
PC0827-100G
PC0827-25G
PC0827-5G
APOSPC0827-100GEA
240
DKK
APOSPC0827-100G
APOSPC0827-25G
APOSPC0827-5G
5-Fluor-2-nitroanisol 99%
5-Fluor-2-nitroanisol
Formel:
C₇H₆FNO₃ Kogepunkt: 272 °C Smeltepunkt: 49 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00077541 CAS-nummer: 448-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...