4-(Pentafluorothio)benzonitrile
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC0804-1G
PC0804-5G
APOSPC0804-1GEA
1750
DKK
APOSPC0804-1G
APOSPC0804-5G
4-(Pentafluorothio)benzonitrile
4-(Pentafluorothio)benzonitrile
Formel:
C₇H₄F₅NS Smeltepunkt: 102…105 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03788521 CAS-nummer: 401892-85-1 UN nummer: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...