Tris(perfluorbutyl)amin for mass spectrometry
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine
PC0568-10X5G PC0568-5G PC0568-5X5G
APOSPC0568-10X5GEA 852 DKK
APOSPC0568-10X5G APOSPC0568-5G APOSPC0568-5X5G
Tris(perfluorbutyl)amin for mass spectrometry
Tris(perfluorbutyl)amin
Formel: C₁₂F₂₇N
Molekylvægt: 671.1 g/mol
Kogepunkt: 178…180 °C
Smeltepunkt: -55
Massefylde: 1.88 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000436
CAS-nummer: 311-89-7
EINECS: 206-223-1