1-Chlor-2,4-dibrom-6-Fluorbenzen 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Chloro-3,5-dibromo-1-fluorobenzene, 2-Chloro-1,5-dibromo-3-fluorobenzene, 1,5-Dibromo-2-chloro-3-fluorobenzene

PC0437-100G PC0437-25G
APOSPC0437-100GEA 2340 DKK
APOSPC0437-100G APOSPC0437-25G
1-Chlor-2,4-dibrom-6-Fluorbenzen 98%
1-Chlor-2,4-dibrom-6-Fluorbenzen
Formel: C₆H₂Br₂ClF
Molekylvægt: 288.34 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00142891
CAS-nummer: 202925-04-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR