5-Fluor-2-methoxybenzoesyre

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC0292-1G PC0292-25G PC0292-5G
APOSPC0292-1GEA 164 DKK
APOSPC0292-1G APOSPC0292-25G APOSPC0292-5G
5-Fluor-2-methoxybenzoesyre
5-Fluor-2-methoxybenzoesyre
Formel: C₈H₇FO₃
Molekylvægt: 170.14 g/mol
Smeltepunkt: 86…90 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00671765
CAS-nummer: 394-04-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR