(Perfluorocyclohexyl)methyl acrylate 90%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC0277-5G PC0277-1G
APOSPC0277-5GEA 2280 DKK
APOSPC0277-5G APOSPC0277-1G
(Perfluorocyclohexyl)methyl acrylate 90%
(Perfluorocyclohexyl)methyl acrylate
Formel: C₁₀H₅F₁₁O₂
Molekylvægt: 366.13 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00080614
CAS-nummer: 40677-94-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR