2-Quinolin-4-yl-1-(4-trifluoromethylphenyl)ethanol 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC0197-1G PC0197-25G PC0197-5G
APOSPC0197-1GEA 1300 DKK
APOSPC0197-1G APOSPC0197-25G APOSPC0197-5G
2-Quinolin-4-yl-1-(4-trifluoromethylphenyl)ethanol 95+%
2-Quinolin-4-yl-1-(4-trifluoromethylphenyl)ethanol
Formel: C₁₈H₁₄F₃NO
Smeltepunkt: 141…143 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03424494
CAS-nummer: 493024-38-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR