4-Vinylbenzyl hexafluoroisopropyl ether

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC0184-1G PC0184-5G
APOSPC0184-1GEA 612 DKK
APOSPC0184-1G APOSPC0184-5G
4-Vinylbenzyl hexafluoroisopropyl ether
4-Vinylbenzyl hexafluoroisopropyl ether
Formel: C₁₂H₁₀F₆O
Molekylvægt: 284.2 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00080684
CAS-nummer: 111158-92-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR