2-Chlor-5-(trifluormethyl)anisol 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Chloro-2-methoxy-4-(trifluoromethyl)benzene
PC0182-10G
PC0182-5G
PC0182-1G
APOSPC0182-10GEA
1310
DKK
APOSPC0182-10G
APOSPC0182-5G
APOSPC0182-1G
2-Chlor-5-(trifluormethyl)anisol 98%
2-Chlor-5-(trifluormethyl)anisol
Formel:
C₈H₆ClF₃O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01631470 CAS-nummer: 402-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...