(2-Chloro-6-fluoro-3-methylphenyl)acetic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC0120-1G PC0120-250MG PC0120-5G
APOSPC0120-1GEA 828 DKK
APOSPC0120-1G APOSPC0120-250MG APOSPC0120-5G
(2-Chloro-6-fluoro-3-methylphenyl)acetic acid
(2-Chloro-6-fluoro-3-methylphenyl)acetic acid
Formel: C₉H₈ClFO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01631365
CAS-nummer: 261762-92-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR