3,4-Difluorbenzhydrazid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC0107-1G
PC0107-5G
APOSPC0107-1GEA
164
DKK
APOSPC0107-1G
APOSPC0107-5G
3,4-Difluorbenzhydrazid
3,4-Difluorbenzhydrazid
Formel:
C₇H₆F₂N₂O Smeltepunkt: 137…140 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00672992 CAS-nummer: 229957-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...