2-Chlor-6-Fluoranisol 97+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC0024-1G PC0024-25G PC0024-5G
APOSPC0024-1GEA 164 DKK
APOSPC0024-1G APOSPC0024-25G APOSPC0024-5G
2-Chlor-6-Fluoranisol 97+%
2-Chlor-6-Fluoranisol
Formel: C₇H₆ClFO
Flammepunkt: 69 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01631573
CAS-nummer: 53145-38-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR