2,2,3,3,3-Pentafluoropropyl-p-toluenesulphonate 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC0022-100G PC0022-25G
APOSPC0022-100GEA 5860 DKK
APOSPC0022-100G APOSPC0022-25G
2,2,3,3,3-Pentafluoropropyl-p-toluenesulphonate 98%
Formel: C₁₀H₉F₅O₃S
Kogepunkt: 75 °C/0,75mmHg
Smeltepunkt: 53…55 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02093346
CAS-nummer: 565-42-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR