α-Brom-m-toluonitril

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-(Bromomethyl)benzonitrile, m-(Bromomethyl)benzonitrile

OR9974-100G OR9974-10G OR9974-25G
APOSOR9974-100GEA 1530 DKK
APOSOR9974-100G APOSOR9974-10G APOSOR9974-25G
α-Brom-m-toluonitril
α-Brom-m-toluonitril
Formel: C₈H₆BrN
Kogepunkt: 130 °C/3,8mmHg
Smeltepunkt: 91…96 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001809
CAS-nummer: 28188-41-2
EINECS: 248-890-1
UN nummer: 3261
ADR: 8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR