Piperidin-4-ol 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Piperidinol, 4-Hydroxypiperidine
OR9943-25G
OR9943-100G
OR9943-10G
APOSOR9943-25GEA
164
DKK
APOSOR9943-25G
APOSOR9943-100G
APOSOR9943-10G
Piperidin-4-ol 95%
Piperidin-4-ol
Formel:
C₅H₁₁NO Kogepunkt: 108…114 °C/10mmHg Smeltepunkt: 86…90 °C Flammepunkt: 108 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005999 CAS-nummer: 5382-16-1 EINECS: 226-373-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...