2-Amino-5-Chlorphenol 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Chloro-2-hydroxyaniline
OR9877-100G
OR9877-25G
OR9877-5G
APOSOR9877-100GEA
942
DKK
APOSOR9877-100G
APOSOR9877-25G
APOSOR9877-5G
2-Amino-5-Chlorphenol 97%
2-Amino-5-Chlorphenol
Formel:
C₆H₆ClNO Molekylvægt: 143.57 g/mol Smeltepunkt: 145…155 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02093863 CAS-nummer: 28443-50-7 EINECS: 249-020-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...