2,3-Dihydro-5-oxo-(1H,5H)-benzo[ij]-quinolizine-6-carboxaldehyde 95+%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
5-Oxo-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinoline-6-carboxaldehyde
OR9669-1G
OR9669-5G
OR9669-25G
APOSOR9669-1GEA
1560
DKK
APOSOR9669-1G
APOSOR9669-5G
APOSOR9669-25G
2,3-Dihydro-5-oxo-(1H,5H)-benzo[ij]-quinolizine-6-carboxaldehyde 95+%
2,3-Dihydro-5-oxo-(1H,5H)-benzo[ij]quinolizine-6-carboxaldehyde
Formel:
C₁₃H₁₁NO₂ Molekylvægt: 213.24 g/mol Smeltepunkt: 214…216 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01567808 CAS-nummer: 386715-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...