3,3-Dimethylbutyraldehyd 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3,3-Dimethylbutanal

OR964405-10G OR964405-25G OR964405-100G OR964405-1G OR964405-5G
APOSOR964405-10GEA 0 DKK
APOSOR964405-10G APOSOR964405-25G APOSOR964405-100G APOSOR964405-1G APOSOR964405-5G
3,3-Dimethylbutyraldehyd 97%
3,3-Dimethylbutyraldehyd
Formel: C₆H₁₂O
Molekylvægt: 100.16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00042807
CAS-nummer: 2987-16-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR