3-(4-Cyanophenyl)propanoic acid 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR964275-1G OR964275-5G
APOSOR964275-1GEA 183 DKK
APOSOR964275-1G APOSOR964275-5G
3-(4-Cyanophenyl)propanoic acid 98%
3-(4-Cyanophenyl)propanoic acid
Formel: C₁₀H₉NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08234823
CAS-nummer: 42287-94-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR